missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éthyle 5-fluoroindole-2-carboxylate 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 150°C |
| Color | Blanc-jaune |
| Quantity | 5 g |
| Formula Weight | 207.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Ethyl 5-Fluoroindole-2-carboxylate |
Sécurité et manipulation
| TSCA | No |