missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Fluoroindole-2-carboxylate 98.0+%, TCI America™

Catalog No. E10351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10351G 1 g
E10355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10351G Supplier TCI America Supplier No. E10351G

Chemical Identifiers

CAS 348-36-7
Molecular Formula C11H10FNO2
Molecular Weight (g/mol) 207.20
MDL Number MFCD00152076
InChI Key VIKOQTQMWBKMNA-UHFFFAOYSA-N
Synonym ethyl 5-fluoroindole-2-carboxylate, 5-fluoroindole-2-carboxylic acid ethyl ester, 2-carbethoxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro-, ethyl ester, 5-fluoro-1h-indole-2-carboxylic acid ethyl ester, ethyl5-fluoro-indole-2-carboxylate, pubchem7276, maybridge1_006190, acmc-1aex1, ksc573a9h
PubChem CID 2727899
IUPAC Name ethyl 5-fluoro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(F)=CC=C2N1

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 1 g
Formula Weight 207.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Fluoroindole-2-carboxylate