missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-fluoroindole-2-carboxylate, 98%

Catalog No. AAL1401003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1401003 1 g
AAL1401006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1401003 Supplier Thermo Scientific Chemicals Supplier No. L1401003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 348-36-7 | C11H10FNO2 | 207.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-36-7
Molecular Formula C11H10FNO2
Molecular Weight (g/mol) 207.20
MDL Number MFCD00152076
InChI Key VIKOQTQMWBKMNA-UHFFFAOYSA-N
Synonym ethyl 5-fluoroindole-2-carboxylate, 5-fluoroindole-2-carboxylic acid ethyl ester, 2-carbethoxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro-, ethyl ester, 5-fluoro-1h-indole-2-carboxylic acid ethyl ester, ethyl5-fluoro-indole-2-carboxylate, pubchem7276, maybridge1_006190, acmc-1aex1, ksc573a9h
PubChem CID 2727899
IUPAC Name ethyl 5-fluoro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(F)=CC=C2N1

Specifications

Melting Point 147°C to 152°C
Quantity 1 g
Beilstein 170254
Formula Weight 207.21
Percent Purity 98%
Chemical Name or Material Ethyl 5-fluoroindole-2-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only