missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-(chlorosulfonyl)-3-furoate, 95%, Thermo Scientific™

Catalog No. p-3629716
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC73303DA 1g Amber Glass Bottle
CC73303DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC73303DA Supplier Thermo Scientific Chemicals Supplier No. CC73303DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 256373-91-8
Molecular Formula C7H7ClO5S
Molecular Weight (g/mol) 238.638
MDL Number MFCD11109333
InChI Key LEKOVVUPHAKDKN-UHFFFAOYSA-N
Synonym ethyl 5-chlorosulfonyl furan-3-carboxylate, ethyl 5-chlorosulfonyl-3-furoate, 3-furancarboxylic acid,5-chlorosulfonyl-, ethyl ester, 5-chlorosulfonyl-furan-3-carboxylic acid ethyl ester, 2-chlorosulfonyl furan-4-carboxylic acid ethyl ester, 3-furancarboxylic acid, 5-chlorosulfonyl-, ethyl ester
PubChem CID 10130959
IUPAC Name ethyl 5-chlorosulfonylfuran-3-carboxylate
SMILES CCOC(=O)C1=COC(=C1)S(=O)(=O)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 238.65
Percent Purity 95%
Chemical Name or Material Ethyl 5-(chlorosulfonyl)-3-furoate