missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-chloropyrazine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. p-7057202
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66630MD 250 mg
AAH6663003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66630MD Supplier Thermo Scientific Chemicals Supplier No. H66630MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54013-04-6
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.60
MDL Number MFCD18070850
InChI Key CFKGDFQQELWOHA-UHFFFAOYSA-N
Synonym ethyl 5-chloropyrazinoate, pyrazinecarboxylic acid, 5-chloro-, ethyl ester, ethyl 5-chloro-2-pyrazinecarboxylate, ethyl5-chloropyrazine-2-carboxylate, 5-chloro-2-pyrazinecarboxylic acid ethyl ester
PubChem CID 465025
IUPAC Name ethyl 5-chloropyrazine-2-carboxylate
SMILES CCOC(=O)C1=CN=C(Cl)C=N1

Specifications

Quantity 250 mg
Beilstein 779320
Formula Weight 186.6
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 5-chloropyrazine-2-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only