missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5-bromothiophene-2-carboxylate, 99%
Description
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses
Solubility
Slightly soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses
Solubility
Slightly soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Boiling Point | 94°C to 96°C (4 mmHg) |
| Quantity | 1 g |
| Solubility Information | Slightly soluble in water |
| Formula Weight | 235.1 |
| Percent Purity | 99% |
| Chemical Name or Material | Ethyl 5-bromothiophene-2-carboxylate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only