missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Bromonicotinate 98.0+%, TCI America™

Catalog No. E08665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08665G 5 g
E086625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08665G Supplier TCI America Supplier No. E08665G

Chemical Identifiers

CAS 20986-40-7
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD00040366
InChI Key PCPIANOJERKFJI-UHFFFAOYSA-N
PubChem CID 140785
IUPAC Name ethyl 5-bromopyridine-3-carboxylate
SMILES CCOC(=O)C1=CC(=CN=C1)Br

Specifications

Melting Point 39°C
Boiling Point 92°C
Quantity 5 g
Synonym ethyl 5-bromonicotinate, 5-bromonicotinic acid ethyl ester, ethyl 5-bromo-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 5-bromo-, ethyl ester, 5-bromo-3-pyridinecarboxylic acid ethyl ester, ethyl5-bromonicotinate, 5-bromo nicotinic acid ethyl ester, 5-bromo-nicotinic acid ethyl ester, ethyl 3-bromo-5-pyridinecarboxylate, 3-bromo-5-pyridine-carboxylic acid ethyl ester
Formula Weight 230.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Bromonicotinate
TSCA No
Recommended Storage Refrigerator