missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromonicotinate, 98%

CAS: 20986-40-7 | C8H8BrNO2 | 230.061 g/mol

Supplier:  Thermo Scientific Chemicals B2078806

 View more versions of this product

Catalog No. AAB2078806


Only null left
Explore available promotions

Description

Description

Ethyl 5-bromonicotinate is used as a key starting material for the preparation of 5-bromonicotinic acid hydrazide via reaction with a hydrazine hydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromonicotinate is used as a key starting material for the preparation of 5-bromonicotinic acid hydrazide via reaction with a hydrazine hydrate.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl 5-bromonicotinate
39°C to 42°C
C8H8BrNO2
5 g
ethyl 5-bromonicotinate, 5-bromonicotinic acid ethyl ester, ethyl 5-bromo-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 5-bromo-, ethyl ester, 5-bromo-3-pyridinecarboxylic acid ethyl ester, ethyl5-bromonicotinate, 5-bromo nicotinic acid ethyl ester, 5-bromo-nicotinic acid ethyl ester, ethyl 3-bromo-5-pyridinecarboxylate, 3-bromo-5-pyridine-carboxylic acid ethyl ester
CCOC(=O)C1=CC(=CN=C1)Br
230.061
230.07
20986-40-7
86°C to 92°C (0.5 mmHg)
MFCD00040366
131473
PCPIANOJERKFJI-UHFFFAOYSA-N
ethyl 5-bromopyridine-3-carboxylate
140785
98%

Chemical Identifiers

20986-40-7
230.061
PCPIANOJERKFJI-UHFFFAOYSA-N
140785
CCOC(=O)C1=CC(=CN=C1)Br
C8H8BrNO2
MFCD00040366
ethyl 5-bromonicotinate, 5-bromonicotinic acid ethyl ester, ethyl 5-bromo-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 5-bromo-, ethyl ester, 5-bromo-3-pyridinecarboxylic acid ethyl ester, ethyl5-bromonicotinate, 5-bromo nicotinic acid ethyl ester, 5-bromo-nicotinic acid ethyl ester, ethyl 3-bromo-5-pyridinecarboxylate, 3-bromo-5-pyridine-carboxylic acid ethyl ester
ethyl 5-bromopyridine-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only