missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromonicotinate, 98%

Catalog No. AAB2078814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2078814 25 g
AAB2078806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2078814 Supplier Thermo Scientific Chemicals Supplier No. B2078814

CAS: 20986-40-7 | C8H8BrNO2 | 230.061 g/mol

Ethyl 5-bromonicotinate is used as a key starting material for the preparation of 5-bromonicotinic acid hydrazide via reaction with a hydrazine hydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromonicotinate is used as a key starting material for the preparation of 5-bromonicotinic acid hydrazide via reaction with a hydrazine hydrate.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 20986-40-7
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD00040366
InChI Key PCPIANOJERKFJI-UHFFFAOYSA-N
Synonym ethyl 5-bromonicotinate, 5-bromonicotinic acid ethyl ester, ethyl 5-bromo-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 5-bromo-, ethyl ester, 5-bromo-3-pyridinecarboxylic acid ethyl ester, ethyl5-bromonicotinate, 5-bromo nicotinic acid ethyl ester, 5-bromo-nicotinic acid ethyl ester, ethyl 3-bromo-5-pyridinecarboxylate, 3-bromo-5-pyridine-carboxylic acid ethyl ester
PubChem CID 140785
IUPAC Name ethyl 5-bromopyridine-3-carboxylate
SMILES CCOC(=O)C1=CC(=CN=C1)Br

Specifications

Melting Point 39°C to 42°C
Boiling Point 86°C to 92°C (0.5 mmHg)
Quantity 25 g
Beilstein 131473
Formula Weight 230.07
Percent Purity 98%
Chemical Name or Material Ethyl 5-bromonicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only