missing translation for 'onlineSavingsMsg'
Learn More

Ethyl(5-bromobenzofuran)-2-carboxylate, 95%, Thermo Scientific™

Catalog No. p-4510461
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC31223CB 250mg
CC31223DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31223CB Supplier Thermo Scientific Chemicals Supplier No. CC31223CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84102-69-2
Molecular Formula C11H9BrO3
Molecular Weight (g/mol) 269.094
InChI Key XLJWAHXKBCDQNP-UHFFFAOYSA-N
Synonym ethyl 5-bromobenzofuran-2-carboxylate, buttpark 34\07-75, 5-bromo-benzofuran-2-carboxylic acid ethyl ester, ethylbromobenzofurancarboxylate, ethyl5-bromobenzofuran-2-carboxylate, ethyl 5-bromo-2-benzofurancarboxylate, ethyl 5-bromo-benzofuran-2-carboxylate, 5-bromo-2-ethoxycarbonyl-1-benzofuran
PubChem CID 735184
IUPAC Name ethyl 5-bromo-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(O1)C=CC(=C2)Br

Specifications

Assay Percent Range 95%
Quantity 250mg
Formula Weight 269.09
Percent Purity 95%
Chemical Name or Material Ethyl (5-bromobenzofuran)-2-carboxylate