missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-benzyloxyindole-2-carboxylate, 97%

Catalog No. p-7044081
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0444003 1 g
AAL0444006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0444003 Supplier Thermo Scientific Chemicals Supplier No. L0444003

CAS: 37033-95-7 | C18H17NO3 | 295.338 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37033-95-7
Molecular Formula C18H17NO3
Molecular Weight (g/mol) 295.338
MDL Number MFCD00022702
InChI Key DCIFXYFKVKDOLL-UHFFFAOYSA-N
Synonym ethyl 5-benzyloxy-1h-indole-2-carboxylate, ethyl 5-benzyloxyindole-2-carboxylate, 5-benzyloxyindole-2-carboxylic acid ethyl ester, 5-benzyloxy-2-carbethoxyindole, ethyl 5-benzyloxy indole-2-carboxylate, 5-benzyloxyindole-2-carboxylic acid ethylester, ethyl 5-phenylmethoxy-1h-indole-2-carboxylate, 5-phenylmethoxy-1h-indole-2-carboxylic acid ethyl ester, 5-benzyloxyindole-2-carboxylic acid*ethyl ester c, 1h-indole-2-carboxylic acid, 5-phenylmethoxy-, ethyl ester
PubChem CID 95721
IUPAC Name ethyl 5-phenylmethoxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)OCC3=CC=CC=C3

Specifications

Melting Point 160°C to 165°C
Quantity 1 g
Beilstein 281129
Formula Weight 295.34
Percent Purity 97%
Chemical Name or Material Ethyl 5-benzyloxyindole-2-carboxylate
EINECSNumber 253-322-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only