missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-aminovalerate hydrochloride, 96%

Catalog No. AAH5168203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5168203 1 g
AAH51682MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5168203 Supplier Thermo Scientific Chemicals Supplier No. H5168203

CAS: 29840-57-1 | C7H16ClNO2 | 181.66 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 29840-57-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD12546205
InChI Key JELXJZASXLHKDV-UHFFFAOYSA-N
Synonym ethyl 5-aminovalerate hydrochloride, ethyl 5-aminopentanoate hydrochloride, ethyl 5-aminovalerate hcl, 5-aminovaleric acid ethyl ester hydrochloride, pentanoic acid, 5-amino-, ethyl ester, hydrochloride, ethyl 5-aminopentanoate hcl, ethyl5-aminovaleratehydrochloride, ethyl 5-azanylpentanoate hydrochloride, 5-amino-valeric acid ethyl ester hydrochloride, 5-aminopentanoic acid ethyl ester hydrochloride
PubChem CID 16094861
IUPAC Name ethyl 5-aminopentanoate;hydrochloride
SMILES CCOC(=O)CCCCN.Cl

Specifications

Linear Formula H2N(CH2)4CO2CH2CH3·HCl
Quantity 1 g
Formula Weight 181.66
Percent Purity 96%
Chemical Name or Material Ethyl 5-aminovalerate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only