missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-(2-furyl)-1-methyl-1H-pyrazole-3-carboxylate, 97%, Thermo Scientific™

Catalog No. CC46923DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC46923DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC46923DA Supplier Thermo Scientific Chemicals Supplier No. CC46923DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104296-35-7
Molecular Formula C11H12N2O3
Molecular Weight (g/mol) 220.228
MDL Number MFCD08271949
InChI Key QEOVLTVAGMDVLQ-UHFFFAOYSA-N
Synonym ethyl 5-furan-2-yl-1-methyl-1h-pyrazole-3-carboxylate, ethyl 5-2-furyl-1-methyl-1h-pyrazole-3-carboxylate, ethyl 5-furan-2-yl-1-methylpyrazole-3-carboxylate, 3-ethoxycarbonyl-5-2-furyl-1-methylpyrazole, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl-1-methyl-, ethyl ester, acmc-20m73c, ethyl 5-2-furyl-1-methylpyrazole-3-carboxylate, 1-methyl-5-2-furyl-1h-pyrazole-3-carboxylic acid ethyl ester, 1h-pyrazole-3-carboxylicacid,5-2-furanyl-1-methyl-,ethyl ester
PubChem CID 7537640
IUPAC Name ethyl 5-(furan-2-yl)-1-methylpyrazole-3-carboxylate
SMILES CCOC(=O)C1=NN(C(=C1)C2=CC=CO2)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 220.23
Percent Purity 97%
Chemical Name or Material Ethyl 5-(2-furyl)-1-methyl-1H-pyrazole-3-carboxylate
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.