missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-pyrazolecarboxylate, 98%, Thermo Scientific™
Specifications
Specifications
| Boiling Point | 138.0°C to 140.0°C (3.0mmHg) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 100mg |
| Formula Weight | 140.14 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl 4-pyrazolecarboxylate |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective glov |
RUO – Research Use Only