missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-pyrazolecarboxylate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 397405000

Quantity 500mg
Packaging Glass bottle

 View more versions of this product

Catalog No. AC397405000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

37622-90-5
140.14
KACZQOKEFKFNDB-UHFFFAOYSA-N
142179
CCOC(=O)C1=CNN=C1
C6H8N2O2
MFCD00010844
ethyl 4-pyrazolecarboxylate, ethyl pyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid ethyl ester, 4-ethoxycarbonylpyrazole, 1h-pyrazole-4-carboxylic acid, ethyl ester, 4-pyrazolecarboxylic acid ethyl ester, ethyl4-pyrazolecarboxylate, zlchem 132, pubchem21939, acmc-209iug
ethyl 1H-pyrazole-4-carboxylate

Specifications

Ethyl 4-pyrazolecarboxylate
138.0°C to 140.0°C (3.0mmHg)
97.5% min. (GC)
C6H8N2O2
MFCD00010844
KACZQOKEFKFNDB-UHFFFAOYSA-N
ethyl 1H-pyrazole-4-carboxylate
142179
98%
37622-90-5
Authentic
Glass bottle
500mg
ethyl 4-pyrazolecarboxylate, ethyl pyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid ethyl ester, 4-ethoxycarbonylpyrazole, 1h-pyrazole-4-carboxylic acid, ethyl ester, 4-pyrazolecarboxylic acid ethyl ester, ethyl4-pyrazolecarboxylate, zlchem 132, pubchem21939, acmc-209iug
CCOC(=O)C1=CNN=C1
140.14
140.14
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective glov

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...