missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-phenylpyrrole-3-carboxylate, 97%

Catalog No. p-7027839
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2487703 1 g
AAB2487706 5 g
AAB2487714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2487703 Supplier Thermo Scientific Chemicals Supplier No. B2487703
Only null left

CAS: 64276-62-6 | C13H13NO2 | 215.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64276-62-6
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD03931521
InChI Key VNZGRYAVCWGZCT-UHFFFAOYSA-N
Synonym ethyl 4-phenylpyrrole-3-carboxylate, 4-phenyl-1h-pyrrole-3-carboxylic acid ethyl ester, 4-phenylpyrrole-3-carboxylic acid ethyl ester, acmc-20akyb, 3-carboethoxy-4-phenylpyrrole, 3-carbethoxy-4-phenylpyrrole, ethyl4-phenyl-1h-pyrrole-3-carboxylate, 1h-pyrrole-3-carboxylicacid, 4-phenyl-, ethyl ester
PubChem CID 4154165
IUPAC Name ethyl 4-phenyl-1H-pyrrole-3-carboxylate
SMILES CCOC(=O)C1=CNC=C1C2=CC=CC=C2

Specifications

Melting Point 126°C to 128°C
Quantity 1 g
Formula Weight 215.25
Percent Purity 97%
Chemical Name or Material Ethyl 4-phenylpyrrole-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only