missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Phenylbutyrate 98.0+%, TCI America™

Catalog No. E10271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10271G 1 g
E10275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10271G Supplier TCI America Supplier No. E10271G

Chemical Identifiers

CAS 10031-93-3
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00026923
InChI Key GGFNXKFGVQQNRV-UHFFFAOYSA-N
Synonym 4-Phenylbutyric Acid Ethyl Ester
PubChem CID 61452
IUPAC Name ethyl 4-phenylbutanoate
SMILES CCOC(=O)CCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 100°C
Quantity 1 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Phenylbutyrate
EINECSNumber (3)-1730
TSCA No