missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Pentenoate 98.0+%, TCI America™

Catalog No. E07155G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07155G Supplier TCI America Supplier No. E07155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1968-40-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00036562
InChI Key PTVSRINJXWDIKP-UHFFFAOYSA-N
Synonym ethyl 4-pentenoate, 4-pentenoic acid ethyl ester, 4-pentenoic acid, ethyl ester, ethyl pent-4-en-1-oate, unii-210um0f8jp, 4-ethoxycarbonylbut-1-ene, pent-4-enoic acid ethyl ester, ethyl allylacetate, ethyl 4-pentenate, ethylpent-4-enoate
PubChem CID 74786
IUPAC Name ethyl pent-4-enoate
SMILES CCOC(=O)CCC=C

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 g
UN Number 3272
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Pentenoate