Learn More
Ethyl 4-pentenoate, 98+%
CAS: 1968-40-7 | C7H12O2 | 128.171 g/mol
Supplier: Thermo Scientific Chemicals A1242206
Description
Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Ethyl 4-pentenoate | |
| 0.9 | |
| 37°C (98°F) | |
| 1.417 | |
| 5 g | |
| 1751643 | |
| Miscible with alcohol. Immiscible with water. | |
| CCOC(=O)CCC=C | |
| 128.171 | |
| 128.17 |
| 1968-40-7 | |
| 146°C | |
| C7H12O2 | |
| MFCD00036562 | |
| UN3272 | |
| ethyl 4-pentenoate, 4-pentenoic acid ethyl ester, 4-pentenoic acid, ethyl ester, ethyl pent-4-en-1-oate, unii-210um0f8jp, 4-ethoxycarbonylbut-1-ene, pent-4-enoic acid ethyl ester, ethyl allylacetate, ethyl 4-pentenate, ethylpent-4-enoate | |
| PTVSRINJXWDIKP-UHFFFAOYSA-N | |
| ethyl pent-4-enoate | |
| 74786 | |
| ≥98% |
Chemical Identifiers
| 1968-40-7 | |
| 128.171 | |
| PTVSRINJXWDIKP-UHFFFAOYSA-N | |
| 74786 | |
| CCOC(=O)CCC=C |
| C7H12O2 | |
| MFCD00036562 | |
| ethyl 4-pentenoate, 4-pentenoic acid ethyl ester, 4-pentenoic acid, ethyl ester, ethyl pent-4-en-1-oate, unii-210um0f8jp, 4-ethoxycarbonylbut-1-ene, pent-4-enoic acid ethyl ester, ethyl allylacetate, ethyl 4-pentenate, ethylpent-4-enoate | |
| ethyl pent-4-enoate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 217-818-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only