missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-nitrophenylglyoxylate, 98+%

Catalog No. AAL0757903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0757903 1 g
AAL0757906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0757903 Supplier Thermo Scientific Chemicals Supplier No. L0757903

CAS: 70091-75-7 | C10H9NO5 | 223.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70091-75-7
Molecular Formula C10H9NO5
Molecular Weight (g/mol) 223.18
MDL Number MFCD00051841
InChI Key ZFCXKZCKJZFZGR-UHFFFAOYSA-N
Synonym ethyl 4-nitrophenylglyoxylate, ethyl 2-4-nitrophenyl-2-oxoacetate, ethyl 4-nitrobenzoylformate, ethyl 4-nitrophenyl oxo acetate, ethyl p-nitrophenylglyoxylate, ethyl-4-nitrophenylglyoxylate, ethyl 4-nitrophenyl oxo acetate #, 4-nitrophenyloxoacetic acid ethyl ester, 4-nitro-phenyl-oxo-acetic acid ethyl ester
PubChem CID 522372
IUPAC Name ethyl 2-(4-nitrophenyl)-2-oxoacetate
SMILES CCOC(=O)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 39°C to 43°C
Boiling Point 152°C to 157°C (0.5 mmHg)
Quantity 1 g
Beilstein 2652331
Formula Weight 223.19
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-nitrophenylglyoxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only