missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Nitrocinnamate 98.0+%, TCI America™

Catalog No. N05985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05985G 5 g
N059825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05985G Supplier TCI America Supplier No. N05985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 953-26-4
Molecular Formula C11H11NO4
Molecular Weight (g/mol) 221.212
MDL Number MFCD00007380
InChI Key PFBQVGXIMLXCQB-VMPITWQZSA-N
Synonym ethyl 4-nitrocinnamate, ethyl p-nitrocinnamate, ethyl 3-4-nitrophenyl acrylate, 2-propenoic acid, 3-4-nitrophenyl-, ethyl ester, cinnamic acid, p-nitro-, ethyl ester, ethyl-4-nitrocinnamate, e-ethyl 3-4-nitrophenyl acrylate, propenoic acid, 3-4-nitrophenyl-, ethyl ester, 4-nitrozimtsaeure-ethylester, ethyl 2e-3-4-nitrophenyl prop-2-enoate
PubChem CID 1268038
IUPAC Name ethyl (E)-3-(4-nitrophenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 138°C
Quantity 5 g
Formula Weight 221.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Nitrocinnamate
RTECSNumber GE0353332
TSCA Yes