missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (4-Nitrobenzoyl)acetate 98.0+%, TCI America™

Catalog No. N051325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N051325G 25 g
N05135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N051325G Supplier TCI America Supplier No. N051325G

Chemical Identifiers

CAS 838-57-3
Molecular Formula C11H11NO5
Molecular Weight (g/mol) 237.211
MDL Number MFCD00007357
InChI Key NGRXSVFCLHVGKU-UHFFFAOYSA-N
Synonym ethyl 4-nitrobenzoylacetate, ethyl 3-4-nitrophenyl-3-oxopropanoate, ethyl p-nitrobenzoyl acetate, ethyl 4-nitrobenzoyl acetate, p-nitrobenzoyl acetic acid ethyl ester, ethyl p-nitrobenzoylacetate, 4-nitrobenzoylacetic acid ethyl ester, ethyl 4-nitro-beta-oxobenzenepropanoate, 4-nitrobenzoyl acetic acid ethyl ester, benzenepropanoic acid, 4-nitro-beta-oxo-, ethyl ester
PubChem CID 13281
IUPAC Name ethyl 3-(4-nitrophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 86°C
Quantity 25 g
Formula Weight 237.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl (4-Nitrobenzoyl)acetate
RTECSNumber AJ1073500
TSCA Yes