missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyloxazole-5-carboxylate, 97+%

Catalog No. AAH5240406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5240406 5 g
AAH5240403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5240406 Supplier Thermo Scientific Chemicals Supplier No. H5240406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20485-39-6 | C7H9NO3 | 155.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20485-39-6
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.15
MDL Number MFCD00062573
InChI Key XNMORZSEENWFLI-UHFFFAOYSA-N
PubChem CID 88558
IUPAC Name ethyl 4-methyl-1,3-oxazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=CO1

Specifications

Quantity 5 g
Synonym ethyl 4-methyloxazole-5-carboxylate, ethyl4-methyloxazole-5-carboxylate, 4-methyloxazole-5-carboxylic acid ethyl ester, 5-ethoxycarbonyl-4-methyloxazole, ethyl 4-methyl-5-oxazolecarboxylate, 5-oxazolecarboxylic acid, 4-methyl-, ethyl ester, 4-methyl-oxazole-5-carboxylic acid ethyl ester, 4-methyl-5-carbethoxyoxazole, ethyl 4-methyl-oxazole-5-carboxylate, ethyl-4-methyl-1,3-oxazole-5-carboxylate
Formula Weight 141.1
Percent Purity ≥97%
Chemical Name or Material Ethyl 4-methyloxazole-5-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-848-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only