missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Imidazolecarboxylate 98.0+%, TCI America™
$50.33 - $149.13
Chemical Identifiers
| CAS | 23785-21-9 |
|---|---|
| Molecular Formula | C6H8N2O2 |
| Molecular Weight (g/mol) | 140.14 |
| MDL Number | MFCD02646639,MFCD07363812 |
| InChI Key | KLWYPRNPRNPORS-UHFFFAOYSA-N |
| Synonym | ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester |
| PubChem CID | 99170 |
| IUPAC Name | ethyl 1H-imidazole-5-carboxylate |
| SMILES | CCOC(=O)C1=CN=CN1 |
Chemical Identifiers
| 23785-21-9 | |
| 140.14 | |
| KLWYPRNPRNPORS-UHFFFAOYSA-N | |
| 99170 | |
| CCOC(=O)C1=CN=CN1 |
| C6H8N2O2 | |
| MFCD02646639,MFCD07363812 | |
| ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester | |
| ethyl 1H-imidazole-5-carboxylate |
Specifications
| 23785-21-9 | |
| White-Yellow | |
| MFCD02646639,MFCD07363812 | |
| ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester | |
| CCOC(=O)C1=CN=CN1 | |
| 140.14 | |
| 140.14 | |
| Crystalline Powder |
| 159°C | |
| C6H8N2O2 | |
| 1 g | |
| KLWYPRNPRNPORS-UHFFFAOYSA-N | |
| ethyl 1H-imidazole-5-carboxylate | |
| 99170 | |
| ≥98.0% (GC,T) | |
| Ethyl 4-Imidazolecarboxylate |
Safety and Handling
TSCA : No