missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Imidazolecarboxylate 98.0+%, TCI America™

Catalog No. E10425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10425G 5 g
E10421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10425G Supplier TCI America Supplier No. E10425G

Chemical Identifiers

CAS 23785-21-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD02646639,MFCD07363812
InChI Key KLWYPRNPRNPORS-UHFFFAOYSA-N
Synonym ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester
PubChem CID 99170
IUPAC Name ethyl 1H-imidazole-5-carboxylate
SMILES CCOC(=O)C1=CN=CN1

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Imidazolecarboxylate