missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydroxy-3-methoxycinnamate 97.0+%, TCI America™

Catalog No. p-7129123
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07395G 5 g
E073925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07395G Supplier TCI America Supplier No. E07395G

Chemical Identifiers

CAS 4046-02-0
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009190
InChI Key ATJVZXXHKSYELS-FNORWQNLSA-N
Synonym ethyl ferulate, ferulic acid ethyl ester, ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, unii-5b8915uelw, ethyl 4'-hydroxy-3'-methoxycinnamate, e-ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, ethyl 2e-3-4-hydroxy-3-methoxyphenyl prop-2-enoate, ethyl 4-hydroxy-3-methoxycinnamate, e-3-4-hydroxy-3-methoxy-phenyl-acrylic acid ethyl ester, ethyl 4-hydroxy-3-methoxycinnamate ferulic acid ethyl ester
PubChem CID 736681
IUPAC Name ethyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 57°C
Color Yellow
Boiling Point 185°C
Quantity 5 g
Formula Weight 222.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Hydroxy-3-methoxycinnamate