missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(Dimethylamino)benzoate 98.0+%, TCI America™

Catalog No. D1744500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1744500G 500 g
D174425G 25 g
D1744100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1744500G Supplier TCI America Supplier No. D1744500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10287-53-3
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD00009115
InChI Key FZUGPQWGEGAKET-UHFFFAOYSA-N
Synonym ethyl 4-dimethylamino benzoate, parbenate, benzoic acid, 4-dimethylamino-, ethyl ester, kayacure epa, ethyl-p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, n,n-dimethylbenzocaine, speedcure edb, ethyl p-dimethylaminobenzoate, unii-829s8d3y0x
PubChem CID 25127
ChEBI CHEBI:52073
IUPAC Name ethyl 4-(dimethylamino)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 66°C
Boiling Point 191°C
Quantity 500 g
Formula Weight 193.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-(Dimethylamino)benzoate
EINECSNumber (3)-3788
TSCA Yes