missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Ethyl 4-(dimethylamino)benzoate

Catalog No. E24905500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E24905500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E24905500G Supplier Sigma Aldrich Supplier No. E24905500G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 4-(dimethylamino)benzoate

Specifications

CAS 10287-53-3
Melting Point 63°C to 66°C
Molecular Formula C11H15NO2
Linear Formula (CH3)2 NC6H4CO2C2H5
Quantity 500 g
MDL Number MFCD00009115
Synonym Parbenate
Molecular Weight (g/mol) 193.24
Percent Purity ≥99%