missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-chlorophenylacetate, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Quantity | 25 g |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Low Melting Solid |
| Assay Percent Range | 98% min. (GC) |
| Molecular Formula | C10H11ClO2 |
| Linear Formula | ClC6H4CH2C(O)OCH2CH3 |
| Molecular Weight (g/mol) | 198.65 |
| Formula Weight | 198.65 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement May cause respiratory irritation. Causes serious eye irritation. Harmful if swallowed. Causes skin irritation. Harmful if inhaled. |
| missing translation for 'healthHazard3' | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse |
| EINECSNumber | 237-906-2 |
RUO – Research Use Only