missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Chloroacetoacetate 95.0+%, TCI America™

Catalog No. p-7124679
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C091125G 25 g
C0911500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C091125G Supplier TCI America Supplier No. C091125G
Only null left

Chemical Identifiers

CAS 638-07-3
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.585
MDL Number MFCD00000939
InChI Key OHLRLMWUFVDREV-UHFFFAOYSA-N
Synonym ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate
PubChem CID 69484
IUPAC Name ethyl 4-chloro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)CCl

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 120°C
Quantity 25 g
UN Number 2922
Formula Weight 164.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Chloroacetoacetate
EINECSNumber (2)-4144
RTECSNumber AK5110000
TSCA Yes