Learn More
Ethyl 4-chloroacetoacetate, 97%
CAS: 638-07-3 | C6H9ClO3 | 164.585 g/mol
Supplier: Thermo Scientific Chemicals A156410B
Description
Ethyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.
Solubility
Miscible with water and most organic solvents.
Notes
Incompatible with strong acids, strong bases, reducing agents and strong oxidizing agents. Store in a cool place.
Specifications
| Ethyl 4-chloroacetoacetate | |
| -8°C | |
| 220°C | |
| C6H9ClO3 | |
| MFCD00000939 | |
| UN2922 | |
| ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate | |
| OHLRLMWUFVDREV-UHFFFAOYSA-N | |
| ethyl 4-chloro-3-oxobutanoate | |
| 69484 | |
| 97% |
| 638-07-3 | |
| 1.212 | |
| 92°C (198°F) | |
| 1.452 | |
| 1000 g | |
| 1761275 | |
| Miscible with water and most organic solvents. | |
| CCOC(=O)CC(=O)CCl | |
| 164.585 | |
| 164.59 |
Chemical Identifiers
| 638-07-3 | |
| 164.585 | |
| OHLRLMWUFVDREV-UHFFFAOYSA-N | |
| 69484 | |
| CCOC(=O)CC(=O)CCl |
| C6H9ClO3 | |
| MFCD00000939 | |
| ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate | |
| ethyl 4-chloro-3-oxobutanoate |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311-H314-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 211-317-0
RTECSNumber : AK5110000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only