missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate, Thermo Scientific Chemicals

Catalog No. AAH5050903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5050903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5050903 Supplier Thermo Scientific Chemicals Supplier No. H5050903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27568-05-4 | Molecular Formula: C13H12ClNO3 | Molecular Weight: 265.69 g/mol

Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate is used as medical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate is used as medical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

Chemical Identifiers

CAS 27568-05-4
Molecular Formula C13H12ClNO3
Molecular Weight (g/mol) 265.69
MDL Number MFCD00173395,MFCD13193759
InChI Key BBOZDELEERNECG-UHFFFAOYSA-N
Synonym ethyl 4-chloro-8-methoxy-3-quinolinecarboxylate, 4-chloro-8-methoxy-quinoline-3-carboxylic acid ethyl ester, 4-chloro-8-methoxyquinoline-3-carboxylic acid ethyl ester, acmc-20a91t, ethyl4-chloro-8-methoxyquinoline-3-carboxylate, ethyl 8-methoxy-4-chloroquinoline-3-carboxylate, ethyl 4-chloro-8-methoxy-quinoline-3-carboxylate, 3-quinolinecarboxylicacid,4-chloro-8-methoxy-,ethyl ester, 4-chloro-8-methoxy-3-quinolinecarboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 4-chloro-8-methoxy-, ethyl ester
PubChem CID 304557
IUPAC Name ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C(OC)=CC=CC2=C1Cl

Specifications

Quantity 1 g
Formula Weight 265.7
Chemical Name or Material Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only