missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Chloro-1-piperidinecarboxylate 98.0+%, TCI America™

Catalog No. E073225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E073225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E073225G Supplier TCI America Supplier No. E073225G
Only null left

Chemical Identifiers

CAS 152820-13-8
Molecular Formula C8H14ClNO2
Molecular Weight (g/mol) 191.655
InChI Key VRTIIQLEEGGJNE-UHFFFAOYSA-N
Synonym 4-Chloro-1-piperidinecarboxylic Acid Ethyl Ester, 1-Carbethoxy-4-chloropiperidine, 4-Chloro-1-(ethoxycarbonyl)piperidine
PubChem CID 44629839
IUPAC Name ethyl 4-chloropiperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(CC1)Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 191.66
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Chloro-1-piperidinecarboxylate