missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromophenylacetate, 99%, Thermo Scientific™

Catalog No. AC392990010
Change view
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392990010 1g Glass bottle
AC392990050 5g Glass bottle
AC392990250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC392990010 Supplier Thermo Scientific Chemicals Supplier No. 392990010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 14062-25-0
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD00016333
InChI Key ZFDCWHPNBWPPHG-UHFFFAOYSA-N
Synonym ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw
PubChem CID 7020609
IUPAC Name ethyl 2-(4-bromophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)Br

Specifications

Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2OC(O)CH2C6H4Br
Quantity 1g
Formula Weight 243.1
Percent Purity 99%
Chemical Name or Material Ethyl 4-bromophenylacetate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only