missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (4-Bromobenzoyl)acetate 98.0+%, TCI America™
$315.72 - $935.64
Chemical Identifiers
| CAS | 26510-95-2 |
|---|---|
| Molecular Formula | C11H11BrO3 |
| Molecular Weight (g/mol) | 271.11 |
| MDL Number | MFCD00231243 |
| InChI Key | PBDYXCKRDRCJDC-UHFFFAOYSA-N |
| Synonym | ethyl 3-4-bromophenyl-3-oxopropanoate, ethyl 4-bromobenzoyl acetate, ethyl 4-bromobenzoylacetate, 3-4-bromo-phenyl-3-oxo-propionic acid ethyl ester, 3-4-bromophenyl-3-oxo-propionic acid ethyl ester, ethyl 3-4-bromophenyl-3-oxo-propionate, benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester, ethyl p-bromobenzoyl acetate, benzenepropanoic acid,4-bromo-b-oxo-, ethyl ester |
| PubChem CID | 2757149 |
| IUPAC Name | ethyl 3-(4-bromophenyl)-3-oxopropanoate |
| SMILES | CCOC(=O)CC(=O)C1=CC=C(C=C1)Br |
Chemical Identifiers
| 26510-95-2 | |
| 271.11 | |
| PBDYXCKRDRCJDC-UHFFFAOYSA-N | |
| 2757149 | |
| CCOC(=O)CC(=O)C1=CC=C(C=C1)Br |
| C11H11BrO3 | |
| MFCD00231243 | |
| ethyl 3-4-bromophenyl-3-oxopropanoate, ethyl 4-bromobenzoyl acetate, ethyl 4-bromobenzoylacetate, 3-4-bromo-phenyl-3-oxo-propionic acid ethyl ester, 3-4-bromophenyl-3-oxo-propionic acid ethyl ester, ethyl 3-4-bromophenyl-3-oxo-propionate, benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester, ethyl p-bromobenzoyl acetate, benzenepropanoic acid,4-bromo-b-oxo-, ethyl ester | |
| ethyl 3-(4-bromophenyl)-3-oxopropanoate |
Specifications
| 26510-95-2 | |
| 124°C | |
| MFCD00231243 | |
| ethyl 3-4-bromophenyl-3-oxopropanoate, ethyl 4-bromobenzoyl acetate, ethyl 4-bromobenzoylacetate, 3-4-bromo-phenyl-3-oxo-propionic acid ethyl ester, 3-4-bromophenyl-3-oxo-propionic acid ethyl ester, ethyl 3-4-bromophenyl-3-oxo-propionate, benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester, ethyl p-bromobenzoyl acetate, benzenepropanoic acid,4-bromo-b-oxo-, ethyl ester | |
| CCOC(=O)CC(=O)C1=CC=C(C=C1)Br | |
| 271.11 | |
| 271.11 | |
| Liquid |
| Yellow | |
| C11H11BrO3 | |
| 1 g | |
| PBDYXCKRDRCJDC-UHFFFAOYSA-N | |
| ethyl 3-(4-bromophenyl)-3-oxopropanoate | |
| 2757149 | |
| ≥98.0% (GC) | |
| Ethyl (4-Bromobenzoyl)acetate |
Safety and Handling
TSCA : No