missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Amino-1-piperidinecarboxylate 97.0+%, TCI America™

Catalog No. A15955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15955G 5 g
A159525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15955G Supplier TCI America Supplier No. A15955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58859-46-4
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.228
MDL Number MFCD00006484
InChI Key GQQQULCEHJQUJT-UHFFFAOYSA-N
Synonym ethyl 4-amino-1-piperidinecarboxylate, 4-amino-1-carbethoxypiperidine, unii-54815srp4z, 1-piperidinecarboxylic acid, 4-amino-, ethyl ester, ethyl 4-aminopiperidinecarboxylate, ethyl-4-amino-1-piperidine carboxylate, 4-amino-1-piperidinecarboxylic acid ethyl ester, ethyl-4-aminopiperidin-1-carboxylat, pubchem2064, acmc-209m7k
PubChem CID 100867
IUPAC Name ethyl 4-aminopiperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(CC1)N

Specifications

Color Yellow
Quantity 5 g
Formula Weight 172.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Amino-1-piperidinecarboxylate