missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 98%

CAS: 2386-26-7 | C11H15NO3 | 209.25 g/mol

Supplier:  Thermo Scientific Chemicals A1736518

 View more versions of this product

Catalog No. AAA1736518


Only null left
Explore available promotions

Description

Description

It is utilized as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is utilized as pharmaceutical intermediate.

Solubility
Slightly soluble in water. 

Notes
Refrigerated (2-4°C). Keep away from strong oxidizing agents and bases.
Specifications

Specifications

Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate
140°C to 143°C
MFCD00022373
3-acetyl-2,4-dimethyl-5-carbethoxypyrrole, 2,4-dimethyl-3-acetyl-5-carbethoxypyrrole, 1h-pyrrole-2-carboxylic acid, 4-acetyl-3,5-dimethyl-, ethyl ester, ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 4-acetyl-3,5-dimethyl-pyrrole-2-carboxylic acid ethyl ester, 3-acetyl-2,4-dimethyl-5-ethoxycarbonyl pyrrole, pyrrole-2-carboxylic acid, 4-acetyl-3,5-dimethyl-, ethyl ester, 4-acetyl-3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, maybridge1_002117, acmc-1cg5g
ALRDOFWBPAZOCW-UHFFFAOYSA-N
ethyl 4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
16947
98%
2386-26-7
C11H15NO3
50 g
Slightly soluble in water.
CCOC(=O)C1=C(C)C(C(C)=O)=C(C)N1
209.25
209.25

Chemical Identifiers

2386-26-7
209.25
ALRDOFWBPAZOCW-UHFFFAOYSA-N
16947
CCOC(=O)C1=C(C)C(C(C)=O)=C(C)N1
C11H15NO3
MFCD00022373
3-acetyl-2,4-dimethyl-5-carbethoxypyrrole, 2,4-dimethyl-3-acetyl-5-carbethoxypyrrole, 1h-pyrrole-2-carboxylic acid, 4-acetyl-3,5-dimethyl-, ethyl ester, ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 4-acetyl-3,5-dimethyl-pyrrole-2-carboxylic acid ethyl ester, 3-acetyl-2,4-dimethyl-5-ethoxycarbonyl pyrrole, pyrrole-2-carboxylic acid, 4-acetyl-3,5-dimethyl-, ethyl ester, 4-acetyl-3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, maybridge1_002117, acmc-1cg5g
ethyl 4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UX9370900

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only