missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 98%

Catalog No. AAA1736530
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1736530 250 g
AAA1736509 10 g
AAA1736518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1736530 Supplier Thermo Scientific Chemicals Supplier No. A1736530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2386-26-7 | C11H15NO3 | 209.25 g/mol

It is utilized as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is utilized as pharmaceutical intermediate.

Solubility
Slightly soluble in water. 

Notes
Refrigerated (2-4°C). Keep away from strong oxidizing agents and bases.

Chemical Identifiers

CAS 2386-26-7
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00022373
InChI Key ALRDOFWBPAZOCW-UHFFFAOYSA-N
Synonym 3-acetyl-2,4-dimethyl-5-carbethoxypyrrole, 2,4-dimethyl-3-acetyl-5-carbethoxypyrrole, 1h-pyrrole-2-carboxylic acid, 4-acetyl-3,5-dimethyl-, ethyl ester, ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 4-acetyl-3,5-dimethyl-pyrrole-2-carboxylic acid ethyl ester, 3-acetyl-2,4-dimethyl-5-ethoxycarbonyl pyrrole, pyrrole-2-carboxylic acid, 4-acetyl-3,5-dimethyl-, ethyl ester, 4-acetyl-3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, maybridge1_002117, acmc-1cg5g
PubChem CID 16947
IUPAC Name ethyl 4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=C(C)C(C(C)=O)=C(C)N1

Specifications

Melting Point 140°C to 143°C
Quantity 250 g
Solubility Information Slightly soluble in water.
Formula Weight 209.25
Percent Purity 98%
Chemical Name or Material Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate
RTECSNumber UX9370900
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only