missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,6-Dihydroxynicotinate 97.0+%, TCI America™

Catalog No. E11101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11101G 1 g
E11105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11101G Supplier TCI America Supplier No. E11101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6975-44-6
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.16
MDL Number MFCD00834976
InChI Key QDHHLXABEXNRJX-UHFFFAOYSA-N
PubChem CID 54676837
IUPAC Name ethyl 6-hydroxy-4-oxo-1,4-dihydropyridine-3-carboxylate
SMILES CCOC(=O)C1=CNC(O)=CC1=O

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 1 g
Synonym ethyl 4,6-dihydroxynicotinate, 4,6-dihydroxynicotinic acid ethyl ester, ethyl 4,6-dihydroxypyridine-3-carboxylate, 4,6-dihydroxypyridine-3-carboxylic acid ethyl ester, 4, 6-dihydroxynicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 4,6-dihydroxy-, ethyl ester, ethyl 6-hydroxy-4-oxo-1h-pyridine-3-carboxylate, ethyl 4-hydroxy-6-oxo-1,6-dihydropyridine-3-carboxylate, 4,6-dihydroxy-nicotinic acid ethyl ester, maybridge3_007431
Formula Weight 183.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4,6-Dihydroxynicotinate