missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,6-dichloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7039798
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5923103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5923103 Supplier Thermo Scientific Chemicals Supplier No. H5923103

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 21168-41-2
Molecular Formula C12H9Cl2NO2
Molecular Weight (g/mol) 270.11
MDL Number MFCD00173348
InChI Key BSSNTDZRVNQGDF-UHFFFAOYSA-N
Synonym ethyl 4,6-dichloro-3-quinolinecarboxylate, 4,6-dichloroquinoline-3-carboxylic acid ethyl ester, ethyl-4,6-dichloroquinoline-3-carboxylate, 3-quinolinecarboxylic acid, 4,6-dichloro-, ethyl ester, ethyl-4, 6-dichloroquinoline-3-carboxylate, 4,6-dichloro-3-quinolinecarboxylic acid ethyl ester, 3-quinolinecarboxylicacid, 4,6-dichloro-, ethyl ester, 4,6-dichloroquinoline-3-carboxylic acid, ethyl ester
PubChem CID 1479087
IUPAC Name ethyl 4,6-dichloroquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=CC(Cl)=CC2=C1Cl

Specifications

Quantity 1 g
Formula Weight 270.11
Percent Purity 97%
Chemical Name or Material Ethyl 4,6-dichloroquinoline-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only