missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4-Dimethyl-3-oxovalerate 98.0+%, TCI America™

Catalog No. p-7126995
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18915ML 5 mL
D189125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18915ML Supplier TCI America Supplier No. D18915ML
Only null left

Chemical Identifiers

CAS 17094-34-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00042886
InChI Key VUYNTIDSHCJIKF-UHFFFAOYSA-N
Synonym ethyl pivaloylacetate, ethyl 4,4,4-trimethylacetoacetate, ethyl 4,4-dimethyl-3-oxovalerate, 4,4-dimethyl-3-oxovaleric acid ethyl ester, pivaloylacetic acid ethyl ester, ethyl 4,4-dimethyl-3-oxo-pentanoate, pentanoic acid, 4,4-dimethyl-3-oxo-, ethyl ester, 4,4-dimethyl-3-oxo-pentanoic acid ethyl ester, ethylpivaloylacetate, pubchem19816
PubChem CID 86950
IUPAC Name ethyl 4,4-dimethyl-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(C)(C)C

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 mL
Formula Weight 172.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4,4-Dimethyl-3-oxovalerate
EINECSNumber (2)-1487&(2)-1505
TSCA Yes