missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4-Dimethyl-3-oxovalerate 98.0+%, TCI America™

Catalog No. d189125ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
D189125ML 25 mL
D18915ML 5 mL
2 options

Catalog No. D189125ML

Supplier: TCI America D189125ML

Only null left

Chemical Identifiers

CAS 17094-34-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00042886
InChI Key VUYNTIDSHCJIKF-UHFFFAOYSA-N
Synonym ethyl pivaloylacetate, ethyl 4,4,4-trimethylacetoacetate, ethyl 4,4-dimethyl-3-oxovalerate, 4,4-dimethyl-3-oxovaleric acid ethyl ester, pivaloylacetic acid ethyl ester, ethyl 4,4-dimethyl-3-oxo-pentanoate, pentanoic acid, 4,4-dimethyl-3-oxo-, ethyl ester, 4,4-dimethyl-3-oxo-pentanoic acid ethyl ester, ethylpivaloylacetate, pubchem19816
PubChem CID 86950
IUPAC Name ethyl 4,4-dimethyl-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(C)(C)C

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 mL
Formula Weight 172.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4,4-Dimethyl-3-oxovalerate
EINECSNumber (2)-1487&(2)-1505
TSCA Yes