missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4-dimethyl-3-oxovalerate, 97%

Catalog No. AAA1975214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1975214 25 g
AAA1975222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1975214 Supplier Thermo Scientific Chemicals Supplier No. A1975214

CAS: 17094-34-7 | C9H16O3 | 172.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17094-34-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00042886
InChI Key VUYNTIDSHCJIKF-UHFFFAOYSA-N
Synonym ethyl pivaloylacetate, ethyl 4,4,4-trimethylacetoacetate, ethyl 4,4-dimethyl-3-oxovalerate, 4,4-dimethyl-3-oxovaleric acid ethyl ester, pivaloylacetic acid ethyl ester, ethyl 4,4-dimethyl-3-oxo-pentanoate, pentanoic acid, 4,4-dimethyl-3-oxo-, ethyl ester, 4,4-dimethyl-3-oxo-pentanoic acid ethyl ester, ethylpivaloylacetate, pubchem19816
PubChem CID 86950
IUPAC Name ethyl 4,4-dimethyl-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(C)(C)C

Specifications

Density 0.967
Boiling Point 88°C to 90°C (10 mmHg)
Quantity 25 g
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl 4,4-dimethyl-3-oxovalerate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 241-162-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only