missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(4-Chloro-2-methylphenoxy)butyrate 98.0+%, TCI America™

Catalog No. E11491G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11491G 1 g
E1149100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11491G Supplier TCI America Supplier No. E11491G
Only null left

Chemical Identifiers

CAS 10443-70-6
Molecular Formula C13H17ClO3
Molecular Weight (g/mol) 256.726
MDL Number MFCD00128924
InChI Key XNKARWLGLZGMGX-UHFFFAOYSA-N
Synonym 4-(4-Chloro-2-methylphenoxy)butyric Acid Ethyl Ester, MCPB Ethyl Ester
PubChem CID 25286
IUPAC Name ethyl 4-(4-chloro-2-methylphenoxy)butanoate
SMILES CCOC(=O)CCCOC1=C(C=C(C=C1)Cl)C

Specifications

Melting Point 3°C
Color Yellow
Quantity 1 g
Physical Form Clear Liquid at 20°C
UN Number 3082
Formula Weight 256.73
Percent Purity ≥98.0% (GC)
Chemical Name or Material Ethyl 4-(4-Chloro-2-methylphenoxy)butyrate
RTECSNumber EK7745000
TSCA No
Recommended Storage Refrigerator