missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4,4,4-Trifluoroacetoacetate 98.0+%, TCI America™
Supplier: TCI America T0810100G
Specifications
| Ethyl 4,4,4-Trifluoroacetoacetate | |
| Colorless | |
| C6H7F3O3 | |
| 100 g | |
| ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester | |
| CCOC(=O)CC(=O)C(F)(F)F | |
| 184.114 | |
| 184.11 | |
| Liquid |
| 372-31-6 | |
| 132°C | |
| MFCD00000424 | |
| 3272 | |
| OCJKUQIPRNZDTK-UHFFFAOYSA-N | |
| ethyl 4,4,4-trifluoro-3-oxobutanoate | |
| 67793 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 372-31-6 | |
| 184.114 | |
| OCJKUQIPRNZDTK-UHFFFAOYSA-N | |
| 67793 | |
| CCOC(=O)CC(=O)C(F)(F)F |
| C6H7F3O3 | |
| MFCD00000424 | |
| ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester | |
| ethyl 4,4,4-trifluoro-3-oxobutanoate |
Safety and Handling
EINECSNumber : (2)-3829
TSCA : Yes