missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-Trifluoroacetoacetate 98.0+%, TCI America™

Catalog No. T081025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T081025G 25 g
T0810100G 100 g
T0810500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T081025G Supplier TCI America Supplier No. T081025G

Chemical Identifiers

CAS 372-31-6
Molecular Formula C6H7F3O3
Molecular Weight (g/mol) 184.114
MDL Number MFCD00000424
InChI Key OCJKUQIPRNZDTK-UHFFFAOYSA-N
Synonym ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester
PubChem CID 67793
IUPAC Name ethyl 4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 g
UN Number 3272
Formula Weight 184.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4,4,4-Trifluoroacetoacetate
EINECSNumber (2)-3829
TSCA Yes