Learn More
Ethyl 3-(trifluoromethyl)crotonate, (E)+(Z), 96%
CAS: 24490-03-7 | C7H9F3O2 | 182.14 g/mol
$344.23 - $344.23
Chemical Identifiers
| CAS | 24490-03-7 |
|---|---|
| Molecular Formula | C7H9F3O2 |
| Molecular Weight (g/mol) | 182.14 |
| MDL Number | MFCD00040846 |
| InChI Key | OSZLARYVWBUKTG-SNAWJCMRSA-N |
| Synonym | ethyl 4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, ethyl 2e-4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, cis + trans, ethyl e-4,4,4-trifluoro-3-methylbut-2-enoate, e-3-trifluoromethyl-2-butenoic acid ethyl ester |
| PubChem CID | 5838607 |
| IUPAC Name | ethyl (E)-4,4,4-trifluoro-3-methylbut-2-enoate |
| SMILES | CCOC(=O)\C=C(/C)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0968804
|
Thermo Scientific Chemicals
L0968804 |
2 g |
Each for $344.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 24490-03-7 | |
| 182.14 | |
| OSZLARYVWBUKTG-SNAWJCMRSA-N | |
| 5838607 | |
| CCOC(=O)\C=C(/C)C(F)(F)F |
| C7H9F3O2 | |
| MFCD00040846 | |
| ethyl 4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, ethyl 2e-4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, cis + trans, ethyl e-4,4,4-trifluoro-3-methylbut-2-enoate, e-3-trifluoromethyl-2-butenoic acid ethyl ester | |
| ethyl (E)-4,4,4-trifluoro-3-methylbut-2-enoate |
Specifications
| 24490-03-7 | |
| 32°C (89°F) | |
| 1.378 | |
| 2 g | |
| 1910081 | |
| OSZLARYVWBUKTG-SNAWJCMRSA-N | |
| ethyl (E)-4,4,4-trifluoro-3-methylbut-2-enoate | |
| 5838607 | |
| 96% |
| 129°C to 130°C | |
| C7H9F3O2 | |
| MFCD00040846 | |
| UN3272 | |
| ethyl 4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, ethyl 2e-4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, cis + trans, ethyl e-4,4,4-trifluoro-3-methylbut-2-enoate, e-3-trifluoromethyl-2-butenoic acid ethyl ester | |
| CCOC(=O)\C=C(/C)C(F)(F)F | |
| 182.14 | |
| 182.14 | |
| Ethyl 3-(trifluoromethyl)crotonate, (E)+(Z) |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only