missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate, 97%

Catalog No. AAA1360906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1360906 5 g
AAA1360914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1360906 Supplier Thermo Scientific Chemicals Supplier No. A1360906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 155377-19-8 | C7H7F3N2O2 | 208.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 155377-19-8
Molecular Formula C7H7F3N2O2
Molecular Weight (g/mol) 208.14
MDL Number MFCD00052083
InChI Key VYXIHSAEOXPAEY-UHFFFAOYSA-N
Synonym ethyl 3-trifluoromethyl pyrazole-4-carboxylate, ethyl 3-trifluoromethyl-1h-pyrazole-4-carboxylate, ethyl 5-trifluoromethyl-1h-pyrazole-4-carboxylate, 3-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 3-trifluoromethyl pyrazole-4-carboxylic acid ethyl ester, ethyl 3-trifluoromethyl-2h-pyrazole-4-carboxylate, 5-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 1h-pyrazole-4-carboxylic acid, 3-trifluoromethyl-, ethyl ester
PubChem CID 596095
IUPAC Name ethyl 5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1)C(F)(F)F

Specifications

Melting Point 142°C to 146°C
Quantity 5 g
Formula Weight 208.14
Percent Purity 97%
Chemical Name or Material Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only