missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Thiophenecarboxylate 95.0+%, TCI America™

Catalog No. E10821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10821G 1 g
E10825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10821G Supplier TCI America Supplier No. E10821G

Chemical Identifiers

CAS 5751-80-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD02179226
InChI Key OYSLMAQEMAJMCL-UHFFFAOYSA-N
Synonym ethyl 3-thenoate, ethyl 3-thiophenecarboxylate, ethylthiophene-3-carboxylate, 3-ethoxycarbonyl thiophene, 3-thiophenecarboxylic acid ethyl ester, 3-thiophenecarboxylic acid, ethyl ester, thiophene-3-carboxylic acid ethyl ester, pubchem16116, ksc493o3n
PubChem CID 79824
IUPAC Name ethyl thiophene-3-carboxylate
SMILES CCOC(=O)C1=CSC=C1

Specifications

Color Yellow
Boiling Point 80°C
Quantity 1 g
Formula Weight 156.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Thiophenecarboxylate