missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Phenylpropionate 98.0+%, TCI America™

Catalog No. p-7135067
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P130425G 25 g
P1304250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P130425G Supplier TCI America Supplier No. P130425G
Only null left

Chemical Identifiers

CAS 2021-28-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00009206
InChI Key JAGZUIGGHGTFHO-UHFFFAOYSA-N
Synonym ethyl 3-phenylpropionate, ethyl hydrocinnamate, ethyl dihydrocinnamate, benzenepropanoic acid, ethyl ester, 3-phenylpropionic acid ethyl ester, hydrocinnamic acid, ethyl ester, ethylhydrocinnamoate, ethyl benzenepropanoate, unii-58e9z9v64d, ethylphenyl propanoate
PubChem CID 16237
IUPAC Name ethyl 3-phenylpropanoate
SMILES CCOC(=O)CCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 248°C
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Phenylpropionate
EINECSNumber (3)-1730
TSCA Yes